Learn More
INSRR Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3055
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487944
  • £377.53 / 100µg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
INSRR Polyclonal antibody AF3055 Specifically detects INSRR in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
INSRR
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human INSRR, Leu24-Leu923, Accession No. P14616
100μg
Primary
3645
Unconjugated
P14616
Goat