missing translation for 'onlineSavingsMsg'
Learn More

IL-4 R alpha Sheep anti-Human, Polyclonal, R&D Systems™

Brand:  RD Systems (Bio-Techne) AF6844

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459904

  • £314.17 / 100µg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

IL-4 R alpha Polyclonal antibody AF6844 Specifically detects IL-4 R alpha in Human samples. It is validated for Immunocytochemistry
Specifications

Specifications

IL-4 R alpha
LYOPH
Immunocytochemistry 5-15 ug/mL
P24394
CD124, CD124 antigen, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-4 R alpha in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse IL-4 R is observed, and less than 1% cross-reactivity with recombinant human (rh) IL-5 R alpha , rhIL-5 R beta , rhIL-9 R, rhIL-13 R alpha 1, and rhIL-13 R alpha 2 is observed.
Human
Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL4R
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R alpha Gly24-His232 Accession # P24394
Immunogen affinity purified
RUO
Primary
3566
Sterile PBS to a final concentration of 0.2 mg/mL.
Videos