missing translation for 'onlineSavingsMsg'
Learn More

IL-4 R alpha Sheep anti-Human, Polyclonal, R&D Systems™

Brand:  RD Systems (Bio-Techne) AB-230-NA

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486554

  • £377.53 / 1mg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL&No.x2011;4&No.160;Rα, Gly24-His232, Accession No. P24394

Applications:

Neutralization, Western Blotting

Specifications

Specifications

IL-4 R alpha
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Purified
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL&No.x2011;4&No.160;Rα, Gly24-His232, Accession No. P24394
1mg
Primary
3566
Unconjugated
P24394
Goat
IgG
RUO
Polyclonal
Human
Videos