missing translation for 'onlineSavingsMsg'
Learn More

IL-36Ra/IL-1F5 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1275

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466854

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-36Ra/IL-1F5 Polyclonal antibody AF1275 Specifically detects IL-36Ra/IL-1F5 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

IL-36Ra/IL-1F5
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction 100 ug/mL
Q9UBH0
FIL1 delta, IL1F5, IL-1HY1, IL-1L1, IL-1RP3, IL36Ra, IL-36RA, IL36RN, Interleukin 36 Receptor Antagonist, Interleukin-36 Receptor Antagonist
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-36Ra/IL-1F5 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IL-1ra is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL36RN
E. coli-derived recombinant human IL-36Ra/IL-1F5 Val2-Asp155 Accession # Q9UBH0
Antigen Affinity-purified
RUO
Primary
26525
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only