Learn More
IL-32 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3040
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13437944
  • £389.41 / 100µg
Estimated Shipment
27-06-2019
Description
Description
IL-32 Polyclonal antibody AF3040 Specifically detects IL-32 in Human samples. It is validated for Western Blotting
Specifications
Specifications
IL-32
LYOPH
Western Blot 0.1 ug/mL
NP_001012651
IL32, IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4, NK4IL-32delta, TAIF, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-32 in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL32
E. coli-derived recombinant human IL-32 alpha Cys2-Lys131 Accession # NP_001012651
Immunogen affinity purified
RUO
Primary
9235
Reconstitute at 0.2 mg/mL in sterile PBS.