missing translation for 'onlineSavingsMsg'
Learn More

IL-3 R alpha/CD123 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF841

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440265

  • £314.17 / 100µg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-3R alpha/CD123 Polyclonal antibody specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-3R alpha/CD123
LYOPH
Western Blot 0.1 ug/mL
Affinity Purified
IL3RA
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-3 R alpha/CD123
Immunogen affinity purified
RUO
Primary
3563
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3R subunit alpha, IL3RA, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-3 R alpha/CD123 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 2% cross-reactivity with recombinant human (rh) IL-2 sR beta , rhIL-2 sR gamma , and rhIL-6 sR is observed.
Human
Videos

For Research Use Only