missing translation for 'onlineSavingsMsg'
Learn More

IL-26/AK155 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1375

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486904

  • £384.13 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

IL-26/AK155 Polyclonal antibody AF1375 Specifically detects IL-26/AK155 in Human samples. It is validated for Western Blotting
Specifications

Specifications

IL-26/AK155
LYOPH
Western Blot 0.1 ug/mL
Q9NPH9
AK155, AK155IL-26interleukin-26, IL26, interleukin 26, Protein AK155
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human IL-26 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human IL-10 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL26
E. coli-derived recombinant human IL-26 (R&D Systems, Catalog # 1375-IL), Lys22-Gln171, Accession # Q9NPH9
Immunogen affinity purified
RUO
Primary
55801
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only