missing translation for 'onlineSavingsMsg'
Learn More

IL-23 R Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1400

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13456924

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-23R Polyclonal antibody specifically detects IL-23R in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-23R
LYOPH
Western Blot 0.1 ug/mL
Q5VWK5
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-23 R in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse IL-23 R is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL23R
Mouse myeloma cell line NS0-derived recombinant human IL-23 R Gly24-Ile353 Accession # Q5VWK5
Immunogen affinity purified
RUO
Primary
149233
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only