missing translation for 'onlineSavingsMsg'
Learn More

IL-22BP Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1087

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466724

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-22BP Polyclonal antibody AF1087 Specifically detects IL-22BP in Human samples. It is validated for Western Blotting
Specifications

Specifications

IL-22BP/IL22 RA2
LYOPH
Western Blot 0.1 ug/mL
NP_851826
class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-22BP in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human IL-22 R is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL22RA2
Mouse myeloma cell line NS0-derived recombinant human IL-22BP Thr22-Pro231 Accession # NP_851826
Immunogen affinity purified
RUO
Primary
116379
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only