missing translation for 'onlineSavingsMsg'
Learn More

IL-21 R Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF991

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440385

  • £314.17 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-21R Polyclonal antibody specifically detects IL-21R in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF.
Specifications

Specifications

IL-21R
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9HBE5
CD360, CD360 antigen, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-21 R Subunit in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse IL-21 R is observed.
Human
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL21R
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-21 R Subunit Cys20-Pro236 Accession # Q9HBE5
Immunogen affinity purified
RUO
Primary
50615
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only