missing translation for 'onlineSavingsMsg'
Learn More

IL-20 R beta Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1788

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457154

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-20 R beta Polyclonal antibody AF1788 Specifically detects IL-20 R beta in Human samples. It is validated for Western Blotting
Specifications

Specifications

IL-20 R beta/FNDC6
LYOPH
Western Blot 0.1 ug/mL
Q6UXL0
FNDC6, DIRS1, FNDC6, IL20R beta, IL-20R2, IL-20Rb, IL20RB, IL20RB interleukin 20 receptor beta, MGC34923
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-20 R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse IL-20 R beta is observed and less than 2% cross-reactivity with recombinant human (rh) IL-20 R alpha , rhIL-10 R beta , rhIFN-gamma R1, and rhIFN-gamma R2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL20RB
Mouse myeloma cell line NS0-derived recombinant human IL-20 R beta Asp30-Ala230 Accession # Q6UXL0
Immunogen affinity purified
RUO
Primary
53833
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only