missing translation for 'onlineSavingsMsg'
Learn More

IL-18/IL-1F4 Propeptide Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF646

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459764

  • £384.13 / 100µg


Estimated Shipment
29-11-2018
Add to basket

Description

Description

IL-18/IL-1F4 Propeptide Polyclonal antibody AF646 Specifically detects IL-18/IL-1F4 Propeptide in Human samples. It is validated for Western Blotting
Specifications

Specifications

IL-18/IL-1F4
LYOPH
Western Blot 0.1 ug/mL
Affinity Purified
IL18
E. coli-derived recombinant human IL-18/IL-1F4
Immunogen affinity purified
RUO
Primary
3606
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects the pro region of human IL-18/IL-1F4 in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with mature recombinant human IL-18 is observed and less than 1% cross-reactivity with mature recombinant mouse IL-18 and recombinant rat IL-18 is observed.
Human
Videos

For Research Use Only