missing translation for 'onlineSavingsMsg'
Learn More

IL-17F Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1335

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13446884

  • £441.00 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-17F Polyclonal antibody AF1335 Specifically detects IL-17F in Human samples. It is validated for ELISA Capture, Immunocytochemistry, Western Blotting
Specifications

Specifications

IL-17F
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
AAK83350
Cytokine ML-1, IL17F, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-17F in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, rhIL-17D, rhIL-17E, and recombinant mouse IL-17F is observed.
Human, Primate
Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL17F
E. coli-derived recombinant human IL-17F (R&D Systems, Catalog # 1335-IL) Arg31-Gln163 Accession # AAK83350
Antigen Affinity-purified
RUO
Primary
112744
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only