missing translation for 'onlineSavingsMsg'
Learn More

IL-17C Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1234

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486824

  • £382.00 / 100µg


Estimated Shipment
28-06-2018
Add to basket

Description

Description

IL-17C Polyclonal antibody AF1234 Specifically detects IL-17C in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

IL-17C
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Affinity Purified
E. coli-derived recombinant human IL&No.x2011;17C, His19-Val197, Accession No. Q9P0M4
100μg
Primary
27189
Human
Unconjugated
Q9P0M4
Goat
IgG
RUO
Polyclonal
Detects human IL-17C in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) IL-17D and rhIL-17E is observed.
Videos

For Research Use Only