missing translation for 'onlineSavingsMsg'
Learn More

IL-12 R beta 2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1959

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437254

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-12 R beta 2 Polyclonal antibody AF1959 Specifically detects IL-12 R beta 2 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

IL-12 R beta 2
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Q99665
IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse IL-12 R beta 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human IL-12 R beta 1 is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL12RB2
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Cys28-Asn622 Accession # Q99665
Immunogen affinity purified
RUO
Primary
3595
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only