missing translation for 'onlineSavingsMsg'
Learn More

IL-1 RAcP/IL-1 R3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF676

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449874

  • £449.92 / 100µg

Estimated Shipment date
from Supplier 06-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-1 RAcP/IL-1 R3 Polyclonal antibody AF676 Specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

IL-1 RAcP/IL-1 R3
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9NPH3
C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 R3, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, IL1RAcP, IL-1RAcP, IL1RAP, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-1 RAcP/IL-1 R3 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse IL-1 RAcP is observed.
Human
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL1RAP
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RAcP/IL-1 R3 Ser21-Glu359 Accession # Q9NPH3
Antigen Affinity-purified
RUO
Primary
3556
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only