missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG2 Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA183241
Code : LD
Additional Details : Weight : 0.01000kg
Description
Mouse anti Human IgG2 antibody, clone 3C7 is specific for human IgG2 and recognizes an epitope in the Fc region.
Human IgG2 Monoclonal antibody specifically detects Human IgG2 in Human samples. It is validated for ELISA.Specifications
Human IgG2 | |
Monoclonal | |
1 mg/mL | |
PBS with <0.1% sodium azide | |
Human IgG from pooled serum. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA | |
3C7 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Human | |
Liquid |