Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA183240
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody reacts with the gamma-2 specific epitope inside the Fc fragment of human IgG1, and does not cross-react with other human immunoglobulins. Mouse anti Human IgG1 antibody, clone 2C11 reacts with the gamma 2-specific epitope inside the Fc fragment of human IgG1. It does not cross react with other human immunoglobulins.
Human IgG1 Fc Monoclonal antibody specifically detects Human IgG1 Fc in Human samples. It is validated for ELISA.Specifications
Human IgG1 Fc | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
Purified human IgG. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA | |
2C11 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Human | |
Liquid |