Learn More
Rabbit anti-Human IgG (H+L) Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen SA510197
164.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SA5-10197 targets Human IgG (H+L) in IDF and IP applications and shows reactivity with Human samples. The SA5-10197 immunogen is human IgG (heavy and light chains).
Human IgG (H+L) Polyclonal antibody specifically detects Human IgG (H+L) in Human samples. It is validated for Immunodiffusion, Immunoprecipitation.Specifications
Human IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Ion-exchange chromatography | |
RUO | |
Human | |
Liquid |
Immunodiffusion, Immunoprecipitation | |
7.4 mg/mL | |
PBS with no preservative; pH 7.8 | |
human IgG (heavy and light chains) | |
20 mg | |
Secondary | |
-20°C | |
IgG |