missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A18818
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Human IgG Fc Polyclonal antibody specifically detects Human IgG Fc in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
Human IgG Fc | |
Polyclonal | |
FITC | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Gamma Immunoglobin Fc region | |
2 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |