missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgA (Heavy chain) Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA511208
Code : LE
Additional Details : Weight : 0.01000kg
Description
By Western blot, MA5-11208 detects Human IgA (alpha) heavy chain at ∽55kD in human, but not mouse samples. By ELISA and WB, MA5-11208 does not react with other human Ig isotypes.
Human IgA (Heavy chain) Monoclonal antibody specifically detects Human IgA (Heavy chain) in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Human IgA (Heavy chain) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
GA01 | |
Unconjugated | |
alpha heavy chain, immunoglobulin alpha chain, IgA heavy chain | |
Purified human alpha heavy chain | |
500 μL | |
Secondary | |
4° C | |
IgG1 κ |