Learn More
F(ab')2-Goat anti-Human Kappa Light Chain Secondary Antibody, HRP, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA174407
301.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 1.24740kg
Description
This antibody has been cross-absorbed with pooled human myeloma proteins with lambda light chains. Purified IgG was prepared from whole serum by affinity chromatography, and F(ab)2 fragments were prepared by pepsin digestion. Goat F(ab')2 anti Human Kappa Light Chain antibody recognizes human kappa light chains. This antibody has been cross absorbed with pooled human myeloma proteins with lambda light chains.
Human Kappa Light Chain Polyclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Human Kappa Light Chain | |
Polyclonal | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.4 | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
HRP | |
Iglc | |
Human kappa light chains. | |
1 mL | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |