Learn More
ICAT Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3016
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13457924
  • £314.17 / 100µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
ICAT Polyclonal antibody AF3016 Specifically detects ICAT in Human samples. It is validated for Western Blotting
Specifications
Specifications
ICAT/CTNNBIP1
LYOPH
Western Blot 0.1 ug/mL
Q9NSA3
beta-catenin-interacting protein ICAT, catenin, beta interacting protein 1, CTNNBIP1, ICATbeta-interacting protein 1, Inhibitor of beta-catenin and Tcf-4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ICAT in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CTNNBIP1
E. coli-derived recombinant human ICAT Met1-Gln81 Accession # Q9NSA3
Immunogen affinity purified
RUO
Primary
56998
Reconstitute at 0.2 mg/mL in sterile PBS.