Learn More
Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2710
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13467734
  • £360.15 / 100µg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Polyclonal antibody AF2710 specifically detects Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q96MM7
EC-1, heparan sulfate 6-O-sulfotransferase 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 in direct ELISAs and Western blots.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
HS6ST2
Mouse myeloma cell line NS0-derived recombinant human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Gln179-Arg605 Accession # Q96MM7
Immunogen affinity purified
RUO
Primary
90161
Reconstitute at 0.2 mg/mL in sterile PBS.