Learn More
GPVI Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3627
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458274
  • £389.41 / 100µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
GPVI Polyclonal antibody AF3627 Specifically detects GPVI in Human samples. It is validated for Western Blotting
Specifications
Specifications
GPVI
LYOPH
Western Blot 0.1 ug/mL
Q9HCN6
Glycoprotein 6, glycoprotein VI (platelet), GP6, GPIV, GPVIplatelet collagen receptor, MGC138168, platelet glycoprotein VI
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human GPVI in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
GP6
Mouse myeloma cell line NS0-derived recombinant human GPVI (R&D Systems, Catalog # 3627-GP) Gln21-Lys267 Accession # Q9HCN6
Immunogen affinity purified
RUO
Primary
51206
Reconstitute at 0.2 mg/mL in sterile PBS.