missing translation for 'onlineSavingsMsg'
Learn More

gp130 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AB-228-NA

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466554

  • £360.15 / 1mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130, Leu24-Glu619 (Glu619Asp), Accession No. P40189

Applications:

Neutralization, Western Blotting

Specifications

Specifications

gp130/CD130
LYOPH
Western Blot 1 ug/mL, Neutralization 20-40 ug/mL
P40189
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Goat
IgG
1 mg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human gp130 in direct ELISAs and Western blots.
Human
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Affinity Purified
IL6ST
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130 Leu24-Glu619 (Glu619Asp) Accession # P40189
Protein A or G purified
RUO
Primary
3572
Reconstitute at 1 mg/mL in sterile PBS.
Videos
Documents