Learn More
GMF-beta Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1276
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13456854
  • £332.22 / 100µg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
GMF-beta Polyclonal antibody AF1276 Specifically detects GMF-beta in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
GMF-beta
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
P60983
glia maturation factor beta, glia maturation factor, beta, GMF, GMFB, GMFbeta, GMF-beta
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human GMF-beta in direct ELISAs and Western blots.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
GMFB
E. coli-derived recombinant human GMF-beta Met1-His142 Accession # P60983
Antigen Affinity-purified
RUO
Primary
2764
Reconstitute at 0.2 mg/mL in sterile PBS.