Learn More
Glucosamine (N-acetyl)-6-Sulfatase/GNS Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2484
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487584
  • £382.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Glucosamine (N-acetyl)-6-Sulfatase/GNS Polyclonal antibody AF2484 Specifically detects Glucosamine (N-acetyl)-6-Sulfatase/GNS in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Glucosamine (N-acetyl)-6-Sulfatase/GNS
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human Glucosamine (N-acetyl)-6-Sulfatase/GNS, Val37-Leu552, Accession No. P15586
100μg
Primary
2799
Unconjugated
P15586
Goat