missing translation for 'onlineSavingsMsg'
Learn More

GFR alpha-2/GDNF R alpha-2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF613

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449604

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 28-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GFR alpha-2/GDNF R alpha-2 Polyclonal antibody AF613 Specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for Blocking (Receptor-ligand), Immunohistochemistry, Western Blotting
Specifications

Specifications

GFR alpha-2/GDNF R alpha-2
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 2-6 ug/mL
O00451
GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF R alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR alpha2, GFRa-2, GFRA2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human GFR alpha-2/GDNF R alpha-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 75% cross-reactivity with recombinant mouse GFR alpha-2 is observed and less than 5% cross-reactivity with recombinant human GFR alpha-1 is observed.
Human
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
GFRA2
Mouse myeloma cell line NS0-derived recombinant human GFR alpha-2/GDNF R alpha-2 Ser22-Ser441 Accession # O00451
Antigen Affinity-purified
RUO
Primary
2675
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only