Learn More
GFI-1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3540
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458224
  • £389.41 / 100µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
GFI-1 Polyclonal antibody AF3540 Specifically detects GFI-1 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
GFI-1
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Q99684
GFI1, GFI-1, growth factor independence-1, growth factor independent 1, growth factor independent 1 transcription repressor, Growth factor independent protein 1, Pal-1, SCN2, Zinc finger protein 163, zinc finger protein Gfi-1, ZNF163, ZNF163FLJ94509
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human GFI-1 in direct ELISAs and Western blots.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
GFI1
E. coli-derived recombinant human GFI-1 Pro2-Leu250 Accession # Q99684
Immunogen affinity purified
RUO
Primary
2672
Reconstitute at 0.2 mg/mL in sterile PBS.