missing translation for 'onlineSavingsMsg'
Learn More

Galectin-2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1153

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436774

  • £314.17 / 100µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Galectin-2 Polyclonal antibody AF1153 Specifically detects Galectin-2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Galectin-2
LYOPH
Western Blot 0.1 ug/mL
P05162
Beta-galactoside-binding lectin L-14-II, Gal-2, GAL2, galectin 2, galectin-2, Galectin2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, LGALS2, MGC75071, S-Lac lectin 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Galectin-2 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human (rh) Galectin-1, rhGalectin-4, rhGalectin-7, and rhGalectin-8 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LGALS2
E. coli-derived recombinant human Galectin-2 (R&D Systems, Catalog # 1153-GA) Met1-Glu132 Accession # P05162
Immunogen affinity purified
RUO
Primary
3957
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only