Learn More
FXR/NR1H4 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3554
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448234
  • £349.86 / 100µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
FXR/NR1H4 Polyclonal antibody AF3554 Specifically detects FXR/NR1H4 in Human samples. It is validated for Western Blotting
Specifications
Specifications
FXR/NR1H4
LYOPH
Western Blot 0.1 ug/mL
Q96RI1
BAR, bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, NR1H4, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14, RIP14RXR-interacting protein 14
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FXR/NR1H4 in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NR1H4
E. coli-derived recombinant human FXR/NR1H4 Leu211-Gln486 Accession # Q96RI1
Immunogen affinity purified
RUO
Primary
9971
Reconstitute at 0.2 mg/mL in sterile PBS.