missing translation for 'onlineSavingsMsg'
Learn More

Furin Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1503

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436974

  • £449.92 / 100µg

Estimated Shipment
11-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Furin Polyclonal antibody AF1503 Specifically detects Furin in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Furin
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P09958
Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FUR, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACE, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PC, PCSK3, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Furin in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant mouse Furin is observed, and less than 1% cross-reactivity with recombinant human (rh) PC-1 and rhPC-2 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FURIN
Mouse myeloma cell line NS0-derived recombinant human Furin Asp108-Glu715 Accession # P09958
Immunogen affinity purified
RUO
Primary
5045
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only