missing translation for 'onlineSavingsMsg'
Learn More

FoxH1 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4248

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448634

  • £449.92 / 100µg

Estimated Shipment date
from Supplier 08-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FoxH1 Polyclonal antibody AF4248 Specifically detects FoxH1 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

FoxH1
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
O75593
Fast-1, FAST1, FAST1forkhead box protein H1, Fast-2, FAST2, Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, forkhead activin signal transducer-1, forkhead box H1, hFAST-1, TGF-beta/activin signal transducer
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FoxH1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human FoxB2 is observed.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FOXH1
E. coli-derived recombinant human FoxH1 Ser164-Ser272 Accession # O75593
Immunogen affinity purified
RUO
Primary
8928
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only