missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Follistatin-like 4/FSTL4 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Brand: RD Systems (Bio-Techne) AF4890
Code : Z1
Additional Details : Weight : 0.00010kg
Description
Follistatin-like 4/FSTL4 Polyclonal antibody AF4890 Specifically detects Follistatin-like 4/FSTL4 in Human samples. It is validated for Western BlottingSpecifications
Follistatin-like 4/FSTL4 | |
LYOPH | |
Western Blot 0.1 ug/mL | |
NP_055897 | |
Follistatin like 4, follistatin-like 4, Follistatin-like protein 4, follistatin-related protein 4, FSL4, FSTL4, KIAA1061 | |
Sheep | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human Follistatin-like 4/FSTL4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse FSTL4 is observed and less than 1% cross-reactivity with recombinant human (rh) FSTL1 and rhFSTL5 is observed. | |
Human |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
FSTL4 | |
Chinese hamster ovary cell line CHO-derived recombinant human Follistatin-like 4/FSTL4 Trp23-Val842 Accession # NP_055897 | |
Immunogen affinity purified | |
RUO | |
Primary | |
23105 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
For Research Use Only