missing translation for 'onlineSavingsMsg'
Learn More

FLRT3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2795

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437794

  • £314.17 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FLRT3 Polyclonal antibody AF2795 Specifically detects FLRT3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

FLRT3
LYOPH
Western Blot 0.1 ug/mL
Q9NZU0
fibronectin leucine rich transmembrane protein 3, Fibronectin-like domain-containing leucine-rich transmembrane protein 3, HH21, KIAA1469, leucine-rich repeat transmembrane protein FLRT3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FLRT3 in direct ELISAs and Western blots. In these formats, approximately 10% cross-reactivity with recombinant human (rh) FLRT1 is observed and 5% cross-reactivity with rhFLRT2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FLRT3
Mouse myeloma cell line NS0-derived recombinant human FLRT3 (R&D Systems, Catalog # 2795-FL) Lys29-Pro528 Accession # Q9NZU0
Antigen Affinity-purified
RUO
Primary
23767
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only