missing translation for 'onlineSavingsMsg'
Learn More

FLRT1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2794

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13427794

  • £410.13 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FLRT1 Polyclonal antibody AF2794 Specifically detects FLRT1 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

FLRT1
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9NZU1
fibronectin leucine rich transmembrane protein 1, Fibronectin-like domain-containing leucine-rich transmembrane protein 1, leucine-rich repeat transmembrane protein FLRT1, MGC21624
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FLRT1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) FLRT3 is observed and less than 1% cross-reactivity with rhFLRT2 is observed.
Human
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FLRT1
Mouse myeloma cell line NS0-derived recombinant human FLRT1 Ile21-Pro524 Accession # Q9NZU1
Antigen Affinity-purified
RUO
Primary
23769
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only