missing translation for 'onlineSavingsMsg'
Learn More

FGF-21 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2539

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467624

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FGF-21 Polyclonal antibody AF2539 specifically detects FGF-21 in Human samples. It is validated for Western Blotting
Specifications

Specifications

FGF-21
LYOPH
Western Blot 0.1 ug/mL
Q9NSA1
FGF21, FGF-21, fibroblast growth factor 21
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FGF-21 in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse (rm) FGF-21 is observed and less than 2% cross-reactivity with recombinant human (rh) FGF-3, rhFGF-4, rhFGF-5, rhFGF-6, rhFGF-7, rmFGF-8b, rhFGF-9, rhFGF-10, rhFGF-11, rhFGF-12, rmFGF-15, rhFGF-16, rhFGF-17, rhFGF-18, rhFGF-19, rhFGF-20, rhFGF-23, rhFGF acidic, and rhFGF basic is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FGF21
E. coli-derived recombinant human FGF-21 His29-Ser209 Accession # Q9NSA1
Immunogen affinity purified
RUO
Primary
26291
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only