missing translation for 'onlineSavingsMsg'
Learn More

FGF-19 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF969

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440365

  • £457.32 / 100µg

Estimated Shipment
11-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FGF-19 Polyclonal antibody AF969 Specifically detects FGF-19 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

FGF-19
LYOPH
Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Blockade of Receptor-ligand Interaction
O95750
FGF19, FGF-19, fibroblast growth factor 19
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FGF-19 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) FGF-7, rhFGF-21 and recombinant mouse FGF-15 is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FGF19
E. coli-derived recombinant human FGF-19 Phe27-Lys216 Accession # O95750
Antigen Affinity-purified
RUO
Primary
9965
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only