missing translation for 'onlineSavingsMsg'
Learn More

FGF-13 1B Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1909

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437224

  • £314.17 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FGF-13 1B Polyclonal antibody AF1909 Specifically detects FGF-13 1B in Human samples. It is validated for Western Blotting
Specifications

Specifications

FGF-13
LYOPH
Western Blot 0.1 ug/mL
Q92913-2
FGF13, FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FGF-13 1B in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) FGF-12 is observed and 5% cross-reactivity with rhFGF-11 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FGF13
E. coli-derived recombinant human FGF-13 isoform 1B Ala2-Thr192 Accession # Q92913-2
Antigen Affinity-purified
RUO
Primary
2258
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only