missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RIII (CD16) Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1597

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447034

  • £441.00 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fc gamma RIII (CD16) Polyclonal antibody AF1597 Specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

Fc gamma RIII (CD16)
LYOPH
Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction
O75015
CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human Fc gamma RIIA is observed and less than 2% cross-reactivity with recombinant mouse CD16 is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FCGR3B
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIIB (R&D Systems, Catalog # 1597-FC) Thr20-Gln208 Accession # O75015
Antigen Affinity-purified
RUO
Primary
2214
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only