missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RIIA/CD32a Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1875

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437204

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fc gamma RIIA/CD32a Polyclonal antibody AF1875 Specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Blocking (Receptor-ligand), Flow Cytometry, Western Blotting
Specifications

Specifications

Fc gamma RIIA/CD32a
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready
AAA35827
CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A, FCGR2A1, FcgRIIA, FcRII-a, FCRIIA, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Fc gamma RIIA/CD32a in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Fc gamma RIIB and rhFc gamma RIIIB is observed.
Human
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FCGR2A
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIA/CD32a Ala36-Ile218 Accession # AAA35827
Antigen Affinity-purified
RUO
Primary
2212
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only