missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RI/CD64 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1257

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13456844

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fc gamma RI/CD64 Polyclonal antibody AF1257 Specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

Fc gamma RI/CD64
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
P12314.2
CD64, CD64 antigen, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FCRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Fc gamma RI/CD64 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Fc gamma RI is observed and 10% cross-reactivity with recombinant human (rh) Fc gamma RIIA and rhFc gamma RIIIB is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FCGR1A
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RI/CD64 Gln16-Pro288 Accession # P12314.2
Immunogen affinity purified
RUO
Primary
2209
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only