missing translation for 'onlineSavingsMsg'
Learn More

FAM3A Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2865

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467834

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 23-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FAM3A Polyclonal antibody AF2865 Specifically detects FAM3A in Human samples. It is validated for Western Blotting
Specifications

Specifications

FAM3A
LYOPH
Western Blot 0.1 ug/mL
P98173
2.19, Cytokine-like protein 2-19, DLD, DXS560S, family with sequence similarity 3, member A, member A protein, XAP-7
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FAM3A in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) FAM3B, rhFAM3C, and rhFAM3D is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FAM3A
Mouse myeloma cell line NS0-derived recombinant human FAM3A Phe34-Ser230 Accession # P98173
Antigen Affinity-purified
RUO
Primary
60343
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only