missing translation for 'onlineSavingsMsg'
Learn More

Exostosin-like 3/EXTL3 Lumenal Domain Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2635

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487684

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Exostosin-like 3/EXTL3 Lumenal Domain Polyclonal antibody AF2635 Specifically detects Exostosin-like 3/EXTL3 Lumenal Domain in Human samples. It is validated for Western Blotting
Specifications

Specifications

Exostosin-like 3/EXTL3
LYOPH
Western Blot 0.1 ug/mL
O43909
botv, DKFZp686C2342, EC 2.4.1.223, exostoses (multiple)-like 3, Exostosin like 3, exostosin-like 3, EXTL1L, EXTL3, EXTR1BOTV, EXT-related protein 1, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, Hereditary multiple exostoses gene isolog, KIAA0519, Multiple exostosis-like protein 3, Putative tumor suppressor protein EXTL3, REG, reg receptor, REGR, RPR
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Exostosin-like 3/EXTL3 Lumenal Domain in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EXTL3
Mouse myeloma cell line NS0-derived recombinant human Exostosin-like 3/EXTL3 Lumenal Domain, Thr52-Ile919, Accession # O43909
Immunogen affinity purified
RUO
Primary
2137
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only