Learn More
Enteropeptidase/Enterokinase Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1585
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13467024
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Enteropeptidase/Enterokinase Polyclonal antibody AF1585 Specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Enteropeptidase/Enterokinase
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P98073
EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTK, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7, PRSS7enteropeptidase, Serine protease 7, TMPRSS15, Transmembrane protease serine 15, transmembrane protease, serine 15
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Enteropeptidase/Enterokinase in direct ELISAs and Western blots.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TMPRSS15
Mouse myeloma cell line NS0-derived recombinant human Enteropeptidase/Enterokinase Leu41-His1019 Accession # P98073
Immunogen affinity purified
RUO
Primary
5651
Reconstitute at 0.2 mg/mL in sterile PBS.