missing translation for 'onlineSavingsMsg'
Learn More

ENPP-7/Alk-SMase Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4924

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458984

  • £314.17 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

ENPP-7/Alk-SMase Polyclonal antibody AF4924 Specifically detects ENPP-7/Alk-SMase in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

ENPP-7/Alk-SMase
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q6UWV6
Alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ALK-SMase, EC 3.1.4.12, ectonucleotide pyrophosphatase/phosphodiesterase 7, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, E-NPP 7, ENPP7, Intestinal alkaline sphingomyelinase, MGC50179, NPP7, NPP-7
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ENPP-7/Alk-SMase in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse ENPP-7 is observed, and less than 5% cross-reactivity with recombinant human (rh) ENPP-2 and rhENPP-5 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ENPP7
Mouse myeloma cell line NS0-derived recombinant human ENPP-7/Alk-SMase Ala22-Ser439 Accession # Q6UWV6
Immunogen affinity purified
RUO
Primary
339221
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only