missing translation for 'onlineSavingsMsg'
Learn More

ENPP-2/Autotaxin Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF5255

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459164

  • £377.53 / 100µg


Estimated Shipment
29-11-2018
Add to basket

Description

Description

ENPP-2/Autotaxin Polyclonal antibody AF5255 Specifically detects ENPP-2/Autotaxin in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

ENPP-2/Autotaxin
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q13822
ATX, ATXFLJ26803, ATX-X, AUTOTAXIN, autotaxin-t, EC 3.1.4.39, ectonucleotide pyrophosphatase/phosphodiesterase 2, ectonucleotide pyrophosphatase/phosphodiesterase family member 2, E-NPP 2, ENPP2, Extracellular lysophospholipase D, Lysophosphatidic Acid, LysoPLD, NPP2, PD-IALPHA, PDNP2, PDNP2NPP2, phosphodiesterase I/nucleotide pyrophosphatase 2, plasma lysophospholipase D
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human ENPP-2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) ENPP-5 and rhENPP-7 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ENPP2
Mouse myeloma cell line NS0-derived recombinant human ENPP-2/Autotaxin, Asp49-Ile863, Accession # Q13822
Antigen Affinity-purified
RUO
Primary
5168
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only