Learn More
EGR1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2818
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13447804
  • £362.60 / 100µg
Estimated Shipment
10-08-2020
Description
Description
EGR1 Polyclonal antibody AF2818 Specifically detects EGR1 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
EGR1
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
P18146
AT225Transcription factor ETR103, early growth response 1, early growth response protein 1, EGR-1, G0S30, KROX24, KROX-24, Nerve growth factor-induced protein A, NGFIA, NGFI-ATranscription factor Zif268, TIS8, ZIF-268, Zinc finger protein Krox-24, ZNF225, ZNF225Zinc finger protein 225
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human EGR1 in direct ELISAs and Western blots.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EGR1
E. coli-derived recombinant human EGR1 Gln282-Ser433 Accession # P18146
Antigen Affinity-purified
RUO
Primary
1958
Reconstitute at 0.2 mg/mL in sterile PBS.