missing translation for 'onlineSavingsMsg'
Learn More

DR3/TNFRSF25 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF943

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440345

  • £457.32 / 100µg

Estimated Shipment date
from Supplier 06-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DR3/TNFRSF25 Polyclonal antibody AF943 Specifically detects DR3/TNFRSF25 in Human samples. It is validated for Western Blotting
Specifications

Specifications

DR3/TNFRSF25
LYOPH
Western Blot 0.1 ug/mL
Q93038
Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), LARD, Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, TNFRSF25, TRAMP, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1, WSL-1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human DR3/TNFRSF25 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse DR3 is observed, and less than 1% cross-reactivity with recombinant human (rh) Fas, rhTNF RI, rhDR6, rh4-1BB, rhCD27, rhCD30, rhCD40, rhGITR, rhHVEM, rhTNF RII and rhOPG is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TNFRSF25
Mouse myeloma cell line NS0-derived recombinant human DR3/TNFRSF25 Gln25-Phe201 Accession # Q93038
Immunogen affinity purified
RUO
Primary
8718
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only